missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva DIGE Gel and DIGE Buffer Kit
Tools for increasing the quality, reproducibility, and convenience of 2-D DIGE using Ettan™ DIGE System
Brand: Cytiva 28-9374-51
403.51 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : GE
Additional Details : Weight : 4.99860kg
Includes: Kit: 2 x 125mL anode buffer; 4 x 125mL cathode buffer; 12 tubes agarose sealing solution
Description
- DIGE Gel run with DIGE Buffer Kit results in spot maps with reproducibility compared to homecast polyacrylamide gels, thereby reducing technical variation between gels
- Ready-to-use
- Comes with long shelf-life
- Eliminates the need to make buffers, cast gels and wash glass plates
- Offers quality and performance of the Laemmli system with reproducibility, convenience and time savings
- Should be used with the DIGE Buffer kit
- 12.5% precast low fluorscent polyacrylamide gel
- Cast in low fluorescent glass cassette developed for second dimension 2-D electrophoresis
- Solid
- Dimension: 10 x 7.7 x 0.03 in. (255 x 196 x 1mm)
- Designed two-buffer system for running up to twelve DIGE Gels simultaneously
- Contains the amount of reagents necessary for a single run of up to 12 DIGE Gels
- Liquid
Specifications
DIGE Gel is a precast polyacrylamide gel in a low fluorescent glass cassette. | |
Colorless | |
12 to 120 kDa | |
Industrial applications: analytical chemistry, research For differential protein analysis | |
1 Kit: 2 x 125 mL anode buffer; 4 x 125 mL cathode buffer; 12 tubes agarose sealing solution |
Buffer based on piperidinopropionamide (PPA) | |
DIGE Gel and Buffer Kit | |
Solid and Liquid | |
Running Buffers and sealing solution for attaching immobiline™ DryStrip gels (ipg strips) to the top of the Polyacrylamide gel | |
2-8°C |
Safety and Handling
Hazard Statement
Supplemental information
- EUH210-Safety data sheet available on request.
Supplemental information
- UNKNOWN ACUTE-90 per cent of the mixture consists of component(s) of unknown acute toxicity.
- UNKNOWN AQUATIC-Contains 90% of components with unknown hazards to the aquatic environment.