missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva DIGE Gel and DIGE Buffer Kit
Tools for increasing the quality, reproducibility, and convenience of 2-D DIGE using Ettan™ DIGE System
Brand: Cytiva 28-9374-52
306.12 GBP valid until 2024-12-31
Use promo code "21631" to get your promotional price.
Includes: Kit: 2 x 125mL anode buffer; 4 x 125mL cathode buffer; 12 tubes agarose sealing solution
Description
- DIGE Gel run with DIGE Buffer Kit results in spot maps with reproducibility compared to homecast polyacrylamide gels, thereby reducing technical variation between gels
- Ready-to-use
- Comes with long shelf-life
- Eliminates the need to make buffers, cast gels and wash glass plates
- Offers quality and performance of the Laemmli system with reproducibility, convenience and time savings
- Should be used with the DIGE Buffer kit
- 12.5% precast low fluorscent polyacrylamide gel
- Cast in low fluorescent glass cassette developed for second dimension 2-D electrophoresis
- Solid
- Dimension: 10 x 7.7 x 0.03 in. (255 x 196 x 1mm)
- Designed two-buffer system for running up to twelve DIGE Gels simultaneously
- Contains the amount of reagents necessary for a single run of up to 12 DIGE Gels
- Liquid
Specifications
290°C | |
Blue | |
Solid and Liquid | |
Running Buffers and sealing solution for attaching immobiline™ DryStrip gels (ipg strips) to the top of the Polyacrylamide Gel, Anode Buffer, cathode Buffer, 12 tubes Agarose sealing solution | |
92.09472 | |
2 - 8°C |
Specially designed two-buffer system for running up to twelve DIGE gels simultaneously | |
DIGE Gel and Buffer Kit | |
For differential protein analysis | |
18°C | |
1 Kit: 2 x 125 mL anode buffer; 4 x 125 mL cathode buffer; 12 tubes agarose sealing solution |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction