missing translation for 'onlineSavingsMsg'
Learn More

Traceable™ Remote-Monitoring RTD Thermometer with Calibration

The ultra-thin cable on this Remote-Monitoring RTD Thermometer lets you measure within doored equipment.

Brand:  Traceable™ 86460-06

Product Code. 15336880

  • £67.73 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Won't affect door seal—ideal for use in remote measurement applications.
  • For use in remote measurement applications.
  • High-impact, chemical-resistant ABS case.
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory.
  • Traceable™ model number 4230.
  • This meter has a 10 ft. ribbon micro-cable that allows you to place the probe within an area and close the door without affecting the probe or the door seal.
  • Accurately monitor temperature within freezers, water baths, heating blocks, incubators, refrigerators, and other areas.
  • The fast-response 100 Ω platinum RTD probe can be used in liquids, air/gas, or frozen material.

Traceable™ Products

  • Traceable™ Products have an individually-numbered Traceable™ Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA.
  • It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement.
  • Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.

Guaranteed to have at least one year of calibration remaining on certificate.

Specifications

Specifications

NIST
3 m
RTD
10 ft.
199°C
-99°C
-99°C to +199°C
±0.5% of reading 2°C
For Use In Remote Measurement Applications
4230DS
Digi-Sense Calibrated Remote-Monitoring RTD Thermometer
390.2°F
-146.2°F
-146.2°F to +390.2°F
Yes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers