Learn More
Cole-Parmer™ Digi-Sense™ Power Adapter for Temperature Datalogging Meters
Get the most out of your thermocouple meter
Brand:  Cole-Parmer™
Product Code. 11749609
  • £90.90 / Each
Estimated Shipment: 02-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For use with Oakton temperature datalogging meters.
  • Powers the meter with out the need for batteries.
  • If you are a 24-hour lab or heavy use laboratory, you need the meter plugged into an electrical source.
Specifications
Specifications
115/240 V
Temperature Datalogging Meters