Learn More
Digi-Sense™ Precalibrated Data Logging USB Vane Anemometer
Versatile handheld or mountable design for quick air velocity sampling or long term monitoring
Brand: Digi-Sense™ 20250-22
Additional Details : Weight : 0.28440kg Transport : UN number : 3481 Chem class : 9
Includes: data analysis software on CD, one 3.6 V replaceable battery, and NIST-traceable calibration report with data supplied by InnoCal.
Description
- Reliable measurements guaranteed
- Precalibrated to NIST-traceable standards
- User-selectable units of measure eliminate the need for conversions
- Measure air velocities up to 3937 ft/min (20.0 m/s) to cover broad range of applications
- Analyze and graph logged data using included software
- User-defined high and low alarm levels with visual LED indication to bring attention to potential problems
- Articulated screw-on mounting stand provides flexible positioning to fit the application
Services
2 Years
Specifications
Datalogging Vane Anemometer | |
80 to 3837 fpm | |
USB | |
HVAC | |
2.9 cm | |
14.6 cm | |
3.5 cm | |
0°C to 50°C |
±3% +40 ft./min. or 0.2 m/s | |
Blue | |
Datalogging Vane Anemometer with NIST Traceable Calibration | |
1.125 in. | |
5.75 in. | |
One 3.6 V lithium battery | |
32°F to 122°F | |
1.375 in. |