Learn More
Digi-Sense™ Precalibrated Pressure and Flow Meter
Easily determine the performance of your heating and cooling systems
Brand:  Digi-Sense™ 20250-13
Product Code. 15850026
  • £545.00 / Each
Estimated Shipment: 19-05-2025
to see stock.
Explore more special offers
Add to basket
Includes:
pitot tube, two rubber hoses for use with pitot tube, USB cable, software CD, ABS plastic carrying case, one 9 V battery, and NIST-traceable calibration report with data supplied by InnoCal.
Description
Description
  • Reliable measurements guaranteed
  • Precalibrated to NIST-traceable standards
  • Large, multiline backlit LCD
  • Save and recall 99 sample readings per parameter
  • Time clock on Max/Min/Avg provides time reference
Services
2 Years
Specifications
Specifications
HVAC
5 cm
8 in.
32°F to 122°F
1/4 in.
3 in.
±0.3% Full-scale
0 to 0.7252 psi
2 in.
pitot tube, two rubber hoses for use with pitot tube, USB cable, software CD, ABS plastic carrying case, one 9 V battery, and NIST-traceable calibration report with data supplied by InnoCal.
20 cm
0°C to 50°C
6.4 mm hose Barb
7.5 cm
LCD
Digital Manometer
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Digi-Sense™ Precalibrated Pressure and Flow Meter >