Learn More
Digi-Sense™ Precalibrated Professional Infrared Thermometers
Digi-Sense™ Precalibrated Professional infrared Thermometers safely and easily locate hot and cold spots, preventing problems and saving costs
Brand:  Digi-Sense™ 20250-07
 View more versions of this product
Product Code. 15830026
  • £402.00 / Each
Estimated Shipment: 16-05-2025
to see stock.
Explore more special offers
Add to basket
Includes: Type K flexible probe, USB cable, software, tripod, carrying case, one 9 V battery, and NIST-traceable calibration report with data supplied by InnoCal.
Description
Description
  • Reliable measurements
  • Precalibrated to NIST-traceable standards
  • High distance-to-sight (D/S) ratios handle your long-distance measurements
  • External type K input for contact measurements
  • Dual-laser sighting to pinpoint target areas
  • Protective lens cover with magnetic lock
  • Pistol-grip handle is comfortable and easy to use
  • Measure surface temperatures quickly and accurately
  • USB interface for easy data transmission
Services
2 Years
Specifications
Show More
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Digi-Sense™ Precalibrated Professional Infrared Thermometers > Product type: 30:1; Temperature range: -58 to 3002°F