missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Digi-Sense Replacement Calibration Salt
Digi-Sense Replacement Calibration Salt is designed especially for hygrometers.
Brand: Fisherbrand™ 35612-90
This item is not returnable.
View return policy
Description
- Keep your hygrometer running smoothly with these replacement items
Specifications
Magnesium chloride hexahydrate | |
Cl2Mg · 6H2O | |
Solid | |
Hygrometer Calibration | |
Calibration Salt |
7791-18-6 | |
203.30 g/mol | |
20 g | |
Traceable™ Hygrometers |