Learn More
Cole-Parmer™ Digi-Sense Traceable™ Color Frame Four-Channel Timer with Calibration
Meet quality standards and regulatory compliance for ISO, FDA, cGMP, VFC, CAP, CLIA and Joint Commission requirements with this timer
Brand:  Cole-Parmer™ 06449-08
Code : NEW
Additional Details : Weight : 0.09000kg
Product Code. 15306910
  • £33.59 / Each
Estimated Shipment: 02-08-2024
to see stock.
Add to basket
Description
Description
Includes:
  • Three colorful faceplates/frames, built-in stand/pocket clip, magnetic back, two 1.5 V button cell batteries and NIST-traceable calibration certificate with data.
  • Three bright, colorful frames permit personalization, or dedicate a certain frame for a certain timing task
  • Four independent timing channels feature unique audible alarms for simultaneous timing
  • Count up or down from 100 hours for longer duration experiments
  • Memory function returns the display to the programmed time
  • Traceable products are calibrated to NIST-traceable standards and ready to use as soon as you receive it
  • Each product goes through an intense vetting process before it can carry the Traceable name and it includes NIST-traceable certification, saving you both time and money by not having to calibrate separately
  • NIST-traceable calibration also helps you meet quality standards and regulatory compliance for ISO, FDA, cGMP, VFC, CAP, CLIA and Joint Commission requirement
Specifications
Specifications
4-channel Timer
100 hr.
4
NIST-Traceable