missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Digital Cel-Gro Tissue Culture Rotators
The Digital Cel-Gro Tissue Culture Rotator has a brushless no-maintenance motor that provides a gentle rotating motion for optimum results.
Brand: Thermo Scientific™ 88882016
1316.55 GBP valid until 2024-12-15
Use promo code "21658" to get your promotional price.
Requires : Minimum 1 drum order together with Rotator
This item is not returnable. View return policy
Description
The Thermo Scientific™ Digital Cel-Gro Tissue Culture Rotator precisely mixes test tube contents. Achieve optimum culture results for tissue and virus growth, hormone production, and sample testing of viruses and cytotoxicity assays.
- Digital display of Speed and Time
- Speed range: 5 – 70 rpm
- Speed Accuracy: ±1 rpm
- Adjustable rocking Angle: 95°- 180°
- Loading Capacity: 5 kg
- Timer Range: 0 - 9999mins
- Optional multiple size drums offering different capacity of tube rotation
Order Info
Please order minimum 1 drum together with the rotator
Services
2 years
Specifications
5 kg | |
95 to 180° | |
100/240 V | |
CE | |
5-40°F | |
Stainless Steel Platform | |
26 lb. | |
Audible alarm | |
Digital Cel-Gro Tissue Culture Rotator | |
Cell Culture | |
20 to 85% | |
2 Years |
±1 rpm | |
5 to 70 rpm | |
0.2 A | |
EU, UK, CN Plug | |
50/60 Hz | |
383 x 260 x 522 mm (15.1 x 10.2 x 20.6 in.) | |
11.8 kg | |
Digital control | |
Digital | |
Accessories: 88882149, 88882150,88882151, 88882152, 88882153, 88882154, 88882155, 88882156, 88882157 | |
0 to 9999 min. |