Learn More
Thermo Scientific™ PT1000 Temperature Probe
For Use With: Thermo Scientific™ Touch Screen Dry Baths/Block Heaters
Brand: Thermo Scientific™ 88870122
This item is not returnable.
View return policy
Description
Increase lab versatility with the Thermo Scientific™ digital dry baths, which offer a range of configurations with interchangeable modular blocks for a variety of application needs. Designed for basic to advanced applications with a max temperature of 130°C, a timer and available in 1, 2 and 4 block sizes.
Thermo Scientific digital dry baths are available in a range of configurations to hold 1, 2 and 4 interchangeable modular blocks to accommodate a variety of vessels and applications.
- Digital controls and display of time and temperature
- Advanced internal temperature sensing probe for outstanding temperature accuracy and control
- Precise temperature control with PID circuit
- Temperature calibration enables to offset the temperature to desired value
- Timer allows the user to accurately monitor the heating time
- Wide range of interchangeable aluminum alloy heat blocks provide versatility and allow for easy cleaning and disinfecting
- Powder-coated steel body construction ensures durability
- Built-in over-temperature protection device warrants sample and user safety
- Temperature deviation adjustment
- Sample preparation
- Enzyme preservation
- Enzyme-substrate reactions
- DNA amplification
- Electrophoresis gel degeneration
- Serum coagulation
- Unit must be ordered with block(s)
Specifications
Temperature Probe, PT1000 | |
LED | |
130°C |
Thermo Scientific™ Touch Screen Dry Baths/Block Heaters | |
Steel | |
Dry Bath/Block Thermometer |