missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Digitonin, Thermo Scientific Chemicals
146.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
500 mg
5 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 240.0°C |
Ignition Residue | 0.3% max |
Packaging | Glass bottle |
Quantity | 500 mg |
Merck Index | 15,318 |
Specific Rotation Condition | −54.00 (20.00°C c=3,in methanol) |
Specific Rotation | − 54.00 |
Solubility Information | Solubility in water: insoluble. Other solubilities: practically insoluble in chloroform and ether,1gr dissolves in 57mL abs. alcohol,in 220mL 95%,alcohol |
Formula Weight | 1229.33 |
Chemical Name or Material | Digitonin, Approx. 50% (TLC) |
Safety and Handling
Product Identifier
- Digitonin
- Danger
- Acute toxicity Category 3
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H301-Toxic if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'