Learn More
Diglycolic acid, 98%, Thermo Scientific Chemicals
£42.10 - £551.00
Chemical Identifiers
CAS | 110-99-6 |
---|---|
Molecular Formula | C4H6O5 |
Molecular Weight (g/mol) | 134.09 |
MDL Number | MFCD00004309 |
InChI Key | QEVGZEDELICMKH-UHFFFAOYSA-N |
Synonym | diglycolic acid, 2,2'-oxydiacetic acid, oxydiacetic acid, acetic acid, 2,2'-oxybis, oxybisacetic acid, oxydiethanolic acid, diglycolicacid, bis carboxymethyl ether, oxodiacetic acid, 3-oxapentanedioic acid |
PubChem CID | 8088 |
IUPAC Name | 2-(carboxymethoxy)acetic acid |
SMILES | OC(=O)COCC(O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10295870
|
Thermo Scientific Acros
162261000 |
100 g | Plastic bottle |
£42.10
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10544101
|
Thermo Scientific Acros
162265000 |
500 g | Plastic bottle |
£148.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10096003
|
Thermo Scientific Acros
162260025 |
2.5 kg | Plastic bottle |
£551.00
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
110-99-6 | |
134.09 | |
QEVGZEDELICMKH-UHFFFAOYSA-N | |
8088 | |
OC(=O)COCC(O)=O |
C4H6O5 | |
MFCD00004309 | |
diglycolic acid, 2,2'-oxydiacetic acid, oxydiacetic acid, acetic acid, 2,2'-oxybis, oxybisacetic acid, oxydiethanolic acid, diglycolicacid, bis carboxymethyl ether, oxodiacetic acid, 3-oxapentanedioic acid | |
2-(carboxymethoxy)acetic acid |
Specifications
110-99-6 | |
White to Beige | |
C4H6O5 | |
MFCD00004309 | |
diglycolic acid, 2,2'-oxydiacetic acid, oxydiacetic acid, acetic acid, 2,2'-oxybis, oxybisacetic acid, oxydiethanolic acid, diglycolicacid, bis carboxymethyl ether, oxodiacetic acid, 3-oxapentanedioic acid | |
QEVGZEDELICMKH-UHFFFAOYSA-N | |
2-(carboxymethoxy)acetic acid | |
8088 | |
98% | |
Crystalline Powder |
141.0°C to 145.0°C | |
98% | |
O(CH2CO2H)2 | |
03, 234 | |
Solubility in water: soluble. Other solubilities: soluble in ethanol | |
OC(=O)COCC(O)=O | |
134.09 | |
134.09 | |
Authentic | |
Diglycolic acid, 98% |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 203-823-5
missing translation for 'rtecsNumber' : AJ1530000
missing translation for 'tsca' : TSCA
- Diglycolic acid
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only