Learn More
Digoxin, 95%, Thermo Scientific Chemicals
£71.66 - £337.68
Chemical Identifiers
CAS | 20830-75-5 |
---|---|
Molecular Formula | C41H64O14 |
Molecular Weight (g/mol) | 780.94 |
MDL Number | MFCD00003674 |
InChI Key | LTMHDMANZUZIPE-PUGKRICDSA-N |
Synonym | digoxin, 12beta-hydroxydigitoxin, digoxine, lanoxin, lanoxicaps, digosin, digossina, digoxina, digoxinum, lanoxicaps tn |
PubChem CID | 2724385 |
ChEBI | CHEBI:4551 |
IUPAC Name | 3-[(3S,5R,8R,9S,10S,12R,13S,14S,17R)-3-[(2R,4S,5S,6R)-5-[(2S,4S,5S,6R)-5-[(2S,4S,5S,6R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tet |
SMILES | CC1C(C(CC(O1)OC2C(OC(CC2O)OC3C(OC(CC3O)OC4CCC5(C(C4)CCC6C5CC(C7(C6(CCC7C8=CC(=O)OC8)O)C)O)C)C)C)O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10460501
|
Thermo Scientific Acros
228120010 |
1 g | Glass bottle |
£71.66
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10862041
|
Thermo Scientific Acros
228120050 |
5 g | Glass bottle |
£337.68
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
20830-75-5 | |
780.94 | |
LTMHDMANZUZIPE-PUGKRICDSA-N | |
2724385 | |
3-[(3S,5R,8R,9S,10S,12R,13S,14S,17R)-3-[(2R,4S,5S,6R)-5-[(2S,4S,5S,6R)-5-[(2S,4S,5S,6R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tet |
C41H64O14 | |
MFCD00003674 | |
digoxin, 12beta-hydroxydigitoxin, digoxine, lanoxin, lanoxicaps, digosin, digossina, digoxina, digoxinum, lanoxicaps tn | |
CHEBI:4551 | |
CC1C(C(CC(O1)OC2C(OC(CC2O)OC3C(OC(CC3O)OC4CCC5(C(C4)CCC6C5CC(C7(C6(CCC7C8=CC(=O)OC8)O)C)O)C)C)C)O)O |
Specifications
20830-75-5 | |
94% min. (on dried substance) (HPLC) | |
MFCD00003674 | |
digoxin, 12beta-hydroxydigitoxin, digoxine, lanoxin, lanoxicaps, digosin, digossina, digoxina, digoxinum, lanoxicaps tn | |
LTMHDMANZUZIPE-PUGKRICDSA-N | |
3-[(3S,5R,8R,9S,10S,12R,13S,14S,17R)-3-[(2R,4S,5S,6R)-5-[(2S,4S,5S,6R)-5-[(2S,4S,5S,6R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tet | |
2724385 | |
780.94 | |
Digoxin |
248.0°C to 250.0°C | |
C41H64O14 | |
15,3186 | |
Solubility in water: insoluble. Other solubilities: soluble in diluted alcohol,pyridine,mixture of,chloroform and alcohol,almost insoluble in ether,acetone,ethylacetate,,chloroform | |
CC1C(C(CC(O1)OC2C(OC(CC2O)OC3C(OC(CC3O)OC4CCC5(C(C4)CCC6C5CC(C7(C6(CCC7C8=CC(=O)OC8)O)C)O)C)C)C)O)O | |
780.94 | |
CHEBI:4551 | |
95% |
Safety and Handling
GHS H Statement
Fatal if swallowed.
GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
R
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 244-068-1
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.