missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Digoxin, 96%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
Digoxin is used in several cardiac pathologies such as atrial fibrillation and heart failure.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Digoxin is used in several cardiac pathologies such as atrial fibrillation and heart failure.
Solubility
Soluble in dimethyl sulfoxide and methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Digoxin is used in several cardiac pathologies such as atrial fibrillation and heart failure.
Solubility
Soluble in dimethyl sulfoxide and methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | ∼250°C (decomposition) |
Quantity | 5 g |
UN Number | UN2811 |
Merck Index | 14,3167 |
Solubility Information | Soluble in dimethyl sulfoxide and methanol. |
Formula Weight | 780.95 |
Percent Purity | 96% |
Chemical Name or Material | Digoxin |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'