missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dihydro-β-erythroidine hydrobromide, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Dihydro-β-erythroidine hydrobromide is a member of the Erythrina alkaloids, it is a competitive nicotinic acetylcholine receptor antagonist with moderate selectivity for the neuronal α4 receptor subunit (IC50 values are 0.19 and 0.37 μM for α4β4 and α4β2 receptors, respectively). Antagonizes behavioral effects of nicotine in vivo. Dihydro-β-erythroidine blocks excitation of striatal GABAergic neurons, which completely suppress polysynaptic inhibition between striatal cholinergic interneurons. Dihydro-β-erythroidine hydrobromide has antidepressive-like effects in mice (forced swim and mouse suspension). Orally bioavailable
Specifications
Specifications
Quantity | 50 mg |
Formula Weight | 356.26 |
Percent Purity | >98% |
Chemical Name or Material | Dihydro-β-erythroidine hydrobromide |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction