missing translation for 'onlineSavingsMsg'
Learn More
Dihydrogen hexachloroplatinate(IV) hexahydrate, low nitrate, Premion™, 99.95% (metals basis), Pt 37.5% min, Thermo Scientific Chemicals Product Code.: 11300008

Dihydrogen hexachloroplatinate(IV) hexahydrate, low nitrate, Premion™, 99.95% (metals basis), Pt 37.5% min, Thermo Scientific Chemicals

Product Code. 11300008
5 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11300008

Brand: Thermo Scientific Alfa Aesar 042815.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Dihydrogen hexachloroplatinate(IV) hexahydrate is used for the determination of potassium. It is also used for the purification of platinum by treating it with ammonium chloride flowed by heating of ammonium hexachloroplatinate with hydrogen yields elemental platinum. It also finds application in catalysis, electroplating, photography, platinum mirrors, printing for etching of zinc, producing fine color in glass and porcelain, indelible ink and microscopy. It is a precursor for the preparation of other platinum catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18497-13-7
Molecular Formula Cl6H14O6Pt
Molecular Weight (g/mol) 517.89
MDL Number MFCD00149910
InChI Key PIJUVEPNGATXOD-UHFFFAOYSA-J
IUPAC Name platinum(4+) dihydrogen hexahydrate hexachloride
SMILES [H+].[H+].O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Pt+4]

Specifications

Melting Point 60°C (decomposition)
Color Red to Brown
Density 2.430 g/mL
Odor Odorless
Quantity 5 g
UN Number UN2507
Sensitivity Hygroscopic
Merck Index 14,7526
Solubility Information Soluble in water,ethanol,ether,ethyl acetate and acetone. Insoluble in nitric acid.
Formula Weight 517.91 (409.82 Anhydrous)
Percent Purity 99.95%
Grade Premion™
Concentration or Composition (by Analyte or Components) Pt 37.5% min
Physical Form Lumps
Assay (metals basis)
Chemical Name or Material Dihydrogen hexachloroplatinate (IV) hexahydrate, Low nitrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZ20K010 Certificate TypeCertificate of Analysis Date26/12/2024 Catalog Number042815.06,042815.03,042815.14
Lot NumberU20K052 Certificate TypeCertificate of Analysis Date17/09/2024 Catalog Number042815.06,042815.03,042815.14
Lot NumberU27K055 Certificate TypeCertificate of Analysis Date06/09/2024 Catalog Number042815.06,042815.03,042815.14
Lot NumberS14K027 Certificate TypeCertificate of Analysis Date09/07/2024 Catalog Number042815.06,042815.03,042815.14
Lot NumberQ30K002 Certificate TypeCertificate of Analysis Date08/05/2024 Catalog Number042815.06,042815.03,042815.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dihydrogen hexachloroplatinate(IV) hexahydrate, low nitrate, Premion™, 99.95% (metals basis), Pt 37.5% min, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.