missing translation for 'onlineSavingsMsg'
Learn More
Dihydrogen hexachloroplatinate(IV) hydrate, 99.9% (metals basis), Thermo Scientific Chemicals Product Code.: 11371698

Dihydrogen hexachloroplatinate(IV) hydrate, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 11371698
25 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11371698

Brand: Thermo Scientific Alfa Aesar 011051.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For Catalysis, Electroplating, pHotography

Catalysis, electroplating, photography, platinum mirrors, printing for etching of zinc, producing fine color in glass and porcelain, indelible ink, microscopy.Dihydrogen hexachloroplatinate(IV) hexahydrate is used for the determination of potassium. It is also used for the purification of platinum by treating it with ammonium chloride followed by heating of ammonium hexachloroplatinate with hydrogen yields elemental platinum. It also finds application in catalysis, electroplating, photography, platinum mirrors, printing for etching of zinc, producing fine color in glass and porcelain, indelible ink and microscopy. It is a precursor for the preparation of other platinum catalysts. In addition to this, it is also used as an analytical reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26023-84-7
Molecular Formula Cl6H2Pt
Molecular Weight (g/mol) 409.80
MDL Number MFCD00149909
InChI Key ZKOQTCCVSRSECD-UHFFFAOYSA-J
Synonym CPA, Chloroplatinic acid
IUPAC Name dihydrogen hexachloroplatinumtetrakis(ylium)
SMILES [H+].[H+].Cl[Pt+4](Cl)(Cl)(Cl)(Cl)Cl

Specifications

Melting Point 60°C (decomposition)
Color Orange to White
Physical Form Crystalline
Quantity 25 g
Assay Percent Range 99.9% (metals basis)
Solubility Information Soluble in water,ethanol,ether,ethyl acetate and acetone. Insoluble in nitric acid.
Formula Weight 409.82 (Anhydrous)
Odor Odorless
Sensitivity Hygroscopic
Appearance Red-brown crystals or lump
Density 2.43 g/cm3 at 20°C
Chemical Name or Material Dihydrogen hexachloroplatinate (IV) hexahydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN25L009 Certificate TypeCertificate of Analysis Date27/02/2025 Catalog Number011051.06,011051.22,011051.03,011051.14
Lot NumberW27K020 Certificate TypeCertificate of Analysis Date05/10/2024 Catalog Number011051.06,011051.22,011051.03,011051.14
Lot NumberU05K014 Certificate TypeCertificate of Analysis Date22/08/2024 Catalog Number011051.06,011051.22,011051.03,011051.14
Lot NumberN20K020 Certificate TypeCertificate of Analysis Date05/03/2024 Catalog Number011051.06,011051.22,011051.03,011051.14
Lot NumberM26K014 Certificate TypeCertificate of Analysis Date05/02/2024 Catalog Number011051.06,011051.22,011051.03,011051.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dihydrogen hexachloroplatinate(IV) hydrate, 99.9% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.