Learn More
Dihydrogen hexachloroplatinate(IV) hydrate, Premion™, 99.999% (metals basis), Thermo Scientific Chemicals
£182.00 - £706.00
Chemical Identifiers
CAS | 26023-84-7 |
---|---|
Molecular Formula | Cl6H2Pt |
Molecular Weight (g/mol) | 409.80 |
MDL Number | MFCD00149909 |
InChI Key | ZKOQTCCVSRSECD-UHFFFAOYSA-J |
Synonym | Hexachloroplatinic acid hydrate |
IUPAC Name | dihydrogen hexachloroplatinumtetrakis(ylium) |
SMILES | [H+].[H+].Cl[Pt+4](Cl)(Cl)(Cl)(Cl)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11300288
|
Thermo Scientific Alfa Aesar
043696.03 |
1 g |
£182.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11390278
|
Thermo Scientific Alfa Aesar
043696.06 |
5 g |
£706.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Dihydrogen hexachloroplatinate(IV) hydrate is used in the production of platinum metal and its compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
26023-84-7 | |
409.80 | |
ZKOQTCCVSRSECD-UHFFFAOYSA-J | |
dihydrogen hexachloroplatinumtetrakis(ylium) |
Cl6H2Pt | |
MFCD00149909 | |
Hexachloroplatinic acid hydrate | |
[H+].[H+].Cl[Pt+4](Cl)(Cl)(Cl)(Cl)Cl |
Specifications
26023-84-7 | |
Red to Brown | |
Odorless | |
MFCD00149909 | |
Hexachloroplatinic acid hydrate | |
ZKOQTCCVSRSECD-UHFFFAOYSA-J | |
dihydrogen hexachloroplatinumtetrakis(ylium) | |
409.82 (Anhydrous) | |
Dihydrogen hexachloroplatinate(IV) hydrate |
60°C (decomposition) | |
99.999% (metals basis) | |
Cl6H2Pt | |
Hygroscopic | |
Soluble in water,alcohol,acetone,ethyl acetate and ether. | |
[H+].[H+].Cl[Pt+4](Cl)(Cl)(Cl)(Cl)Cl | |
409.80 | |
Crystals |
Safety and Handling
Do not breathe dust. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
P234-P260-P264b-P270-P271-P272-P280-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P390-P501c
H290-H300-H314-H317-H334-H335-H372
missing translation for 'dotInformation' : DOT Class: 8, Packaging Group: III
missing translation for 'einecsNumber' : 241-010-7
missing translation for 'rtecsNumber' : TP1500000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only