missing translation for 'onlineSavingsMsg'
Learn More

Diiodomethane, 99+%, stabilized with silver wire

Product Code. 10183940 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
100g
2.5kg
25g
500g
This item is not returnable. View return policy

Product Code. 10183940

Brand: Thermo Scientific Acros 169831000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 75-11-6 | CH2I2 | 267.84 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-11-6, 7440-22-4
Molecular Formula CH2I2
Molecular Weight (g/mol) 267.84
MDL Number MFCD00001079
InChI Key NZZFYRREKKOMAT-UHFFFAOYSA-N
Synonym methylene iodide, methane, diiodo, methylene diiodide, mi-gee, dijodmethan, methylenjodid, dijodmethan czech, methylenjodid czech, di-iodomethane, diiodo-methane
PubChem CID 6346
IUPAC Name diiodomethane
SMILES ICI

Specifications

Melting Point 6.0°C
Color Red to Yellow
Density 3.3200g/mL
Boiling Point 181.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula CH2I2
Refractive Index 1.737
Quantity 100 g
Beilstein 01,71
Fieser 09,170; 11,182; 17,116
Merck Index 15,6137
Specific Gravity 3.32
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with alcohol,propanol,isopropanol,,hexane,cyclohexane,ether,chloroform and benzene
Stabilizer Silver wire
Viscosity 2.80 mPa.s (20°C)
Formula Weight 267.83
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Diiodomethane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Diiodomethane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Methylene iodide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.