Learn More
Diisobutylaluminium hydride, 1.1M solution in cyclohexane, AcroSeal™, Thermo Scientific Chemicals
£65.12 - £275.37
Specifications
Chemical Name or Material | Diisobutylaluminium hydride |
---|---|
Color | Undesignated |
Name Note | 1.1M Solution in Cyclohexane |
CAS | 1191-15-7, 110-82-7 |
Melting Point | -70.0°C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10266942
|
Thermo Scientific Acros
201031000 |
100 mL | AcroSeal™ Glass bottle |
£65.12
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10747213
|
Thermo Scientific Acros
201038000 |
800 mL | AcroSeal™ Glass Bottle |
£275.37
800mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid, Reduction reagentSpecifications
Diisobutylaluminium hydride | |
1.1M Solution in Cyclohexane | |
-70.0°C | |
[(CH3)2CHCH2]2AIH | |
04, IV, 4400 | |
01,260; 02,140; 03,101; 04,158; 05,224; 06,198; 07,111; 08,173; 09,171; 10,149; 11,185; 12,191; 13,115; 14,138; 15,137; 16,27; 17,123 | |
15, 3212 | |
Solubility in water: vigorous reaction. Other solubilities: miscible with saturated aliphatic and aromatic, hydrocarbons | |
0.78 | |
Solution |
Undesignated | |
1191-15-7, 110-82-7 | |
C8H19Al | |
MFCD00008928 | |
19.5 to 20.5% (DIBAL-H, on Al basis) | |
0.7800g/mL | |
DIBAL-H, 1.1M solution in cyclohexane | |
142.22 | |
−018°C |
Safety and Handling
GHS H Statement:
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.
In contact with water releases flammable gases which may ignite spontaneously.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
Store in a dry place. Store in a closed container.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
- Diisobutylaluminium hydride, 1.1M solution in cyclohexane
Palabra de advertencia
- Danger
Clasificación
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Aspiration toxicity Category 1
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 A
- Specific target organ toxicity Category 3
- Substance which\, in contact with water\, emits flammable gases Category 1
Indicaciones de peligro
- H304-May be fatal if swallowed and enters airways.
- H318-Causes serious eye damage.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H314-Causes severe skin burns and eye damage.
- H336-May cause drowsiness or dizziness.
- H260-In contact with water releases flammable gases which may ignite spontaneously.
- H225-Highly flammable liquid and vapour.
- EUH014-Reacts violently with water.
Consejos de prudencia
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P331-Do NOT induce vomiting.
- P402+P404-Store in a dry place. Store in a closed container.
RUO – Research Use Only