missing translation for 'onlineSavingsMsg'
Learn More

Diisobutylaluminium hydride, 1M solution in hexane, AcroSeal™, Thermo Scientific Chemicals

£61.04 - £282.41

Specifications

Chemical Name or Material Diisobutylaluminium hydride
Name Note 1M Solution in Hexane
CAS 1191-15-7, 110-54-3
Melting Point -70.0°C
Density 0.7010g/mL
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10309812
View Documents
Thermo Scientific Acros
183791000
100 mL AcroSeal™ Glass bottle
£61.04
100mL
Please to purchase this item. Need a web account? Register with us today!
10368003
View Documents
Thermo Scientific Acros
183798000
800 mL AcroSeal™ Glass Bottle
£282.41
800mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid, Reduction reagent
Specifications

Specifications

Diisobutylaluminium hydride
1191-15-7, 110-54-3
0.7010g/mL
C8H19Al
MFCD00008928
01,260; 02,140; 03,101; 04,158; 05,224; 06,198; 07,111; 08,173; 09,171; 10,149; 11,185; 12,191; 13,115; 14,138; 15,137; 16,27; 17,123
0.701
Solubility in water: reacts
142.22
1M Solution in Hexane
-70.0°C
−23°C
[(CH3)2CHCH2]2AlH
04, IV, 4400
15, 3212
DIBAL-H
142.22
Solution
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Diisobutylaluminium hydride, 1M solution in hexane, AcroSeal™, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.