missing translation for 'onlineSavingsMsg'
Learn More
Diisobutylaluminum hydride, 1M solution in hexane, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific™ Product Code.: 15484875

Diisobutylaluminum hydride, 1M solution in hexane, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific™

Product Code. 15484875
50 mL
Click to view available options
Quantity:
50 mL
This item is not returnable. View return policy

Product Code. 15484875

Brand: Thermo Scientific Alfa Aesar 042593.AD

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1191-15-7
MDL Number MFCD00008928

Specifications

Color Colorless
Density 0.701g/mL
Flash Point −23°C (−10°F)
Packaging ChemSeal™ bottles
Linear Formula [(CH3)2CHCH2]2AlH
Quantity 50 mL
UN Number UN3399
Sensitivity Air & Moisture Sensitive
Formula Weight 142.22
Physical Form Liquid
Chemical Name or Material Diisobutylaluminum hydride
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Diisobutylaluminum hydride, 1M solution in hexane, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.