Learn More
Diisobutylamine, 99%, Thermo Scientific Chemicals
£33.87 - £117.59
Chemical Identifiers
CAS | 110-96-3 |
---|---|
Molecular Formula | C8H20N |
Molecular Weight (g/mol) | 130.25 |
MDL Number | MFCD00008930 |
InChI Key | OBYVIBDTOCAXSN-OCAPTIKFSA-O |
Synonym | diisobutylamine, 1-propanamine, 2-methyl-n-2-methylpropyl, bis 2-methylpropyl amine, amine, diisobutyl, bis beta-methylpropyl amine, di-isobutylamine, n,n-bis 2-methylpropyl amine, unii-t18y0a819s, ccris 6232, di-2-methylpropyl amine |
PubChem CID | 8085 |
IUPAC Name | 2-methyl-N-(2-methylpropyl)propan-1-amine |
SMILES | CC[C@H](C)[NH2+][C@H](C)CC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10583131
|
Thermo Scientific Acros
159075000 |
500 mL |
£33.87
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10071740
|
Thermo Scientific Acros
159070010 |
1 L |
£55.98
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10508883
|
Thermo Scientific Acros
159070025 |
2.5 L |
£117.59
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
110-96-3 | |
130.25 | |
OBYVIBDTOCAXSN-OCAPTIKFSA-O | |
8085 | |
CC[C@H](C)[NH2+][C@H](C)CC |
C8H20N | |
MFCD00008930 | |
diisobutylamine, 1-propanamine, 2-methyl-n-2-methylpropyl, bis 2-methylpropyl amine, amine, diisobutyl, bis beta-methylpropyl amine, di-isobutylamine, n,n-bis 2-methylpropyl amine, unii-t18y0a819s, ccris 6232, di-2-methylpropyl amine | |
2-methyl-N-(2-methylpropyl)propan-1-amine |
Specifications
110-96-3 | |
0.7400g/mL | |
26°C | |
98.5% min. (GC) | |
C8H20N | |
1.4071 to 1.4091 | |
04, 166 | |
diisobutylamine, 1-propanamine, 2-methyl-n-2-methylpropyl, bis 2-methylpropyl amine, amine, diisobutyl, bis beta-methylpropyl amine, di-isobutylamine, n,n-bis 2-methylpropyl amine, unii-t18y0a819s, ccris 6232, di-2-methylpropyl amine | |
OBYVIBDTOCAXSN-OCAPTIKFSA-O | |
2-methyl-N-(2-methylpropyl)propan-1-amine | |
8085 | |
129.24 | |
Liquid |
-77°C | |
137°C to 139°C | |
Authentic | |
Glass bottle | |
[(CH3)2CHCH2]2NH | |
MFCD00008930 | |
0.74 | |
Solubility in water: 5g/L water (20°C) | |
CC[C@H](C)[NH2+][C@H](C)CC | |
130.25 | |
0.8 mPa.s (20°C) | |
99% | |
Diisobutylamine |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Toxic if swallowed.
Flammable liquid and vapour.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 203-819-3
- Diisobutylamine
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H301-Toxic if swallowed.
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only