Learn More
Diisopropanolamine, 98+%, sum of isomers, Thermo Scientific Chemicals
£22.70 - £57.40
Chemical Identifiers
CAS | 110-97-4 |
---|---|
Molecular Formula | C6H15NO2 |
Molecular Weight (g/mol) | 133.19 |
MDL Number | MFCD00004531 |
InChI Key | LVTYICIALWPMFW-UHFFFAOYNA-N |
Synonym | diisopropanolamine, 1,1'-iminodipropan-2-ol, bis 2-hydroxypropyl amine, 2-propanol, 1,1'-iminobis, bis 2-propanol amine, 1,1'-iminodi-2-propanol, dipa alcohol, di-2-propanolamine, 1,1'-azanediylbis propan-2-ol, dipropyl-2,2'-dihydroxy-amine |
PubChem CID | 8086 |
IUPAC Name | 1-[(2-hydroxypropyl)amino]propan-2-ol |
SMILES | CC(O)CNCC(C)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11438377
|
Thermo Scientific Alfa Aesar
A18451.30 |
250 g |
£22.70
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11428377
|
Thermo Scientific Alfa Aesar
A18451.0B |
1000 g |
£57.40
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Bis(2-hydroxypropyl)amine (Diisopropanolamine) is used to study its effects upon choline uptake and phospholipid synthesis in Chinese hamster ovary (CHO) cells. It is a versatile chemical that is widely used as an emulsifier, stabilizer, surfactant, and chemical building block. It can neutralize pH, act as a buffer (stabilize pH), or add basicity (alkalinity) to a solution. Major applications include personal care products, metalworking products, and natural gas purification. Diisopropanolamine is also used in coatings, paint strippers, paper, photographic intermediates, plastics, polyurethanes, and textiles. It is frequently used to neutralize fatty acids and sulfonic acidbased surfactants.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBis(2-hydroxypropyl)amine (Diisopropanolamine) is used to study its effects upon choline uptake and phospholipid synthesis in Chinese hamster ovary (CHO) cells. It is a versatile chemical that is widely used as an emulsifier, stabilizer, surfactant, and chemical building block. It can neutralize pH, act as a buffer (stabilize pH), or add basicity (alkalinity) to a solution. Major applications include personal care products, metalworking products, and natural gas purification. Diisopropanolamine is also used in coatings, paint strippers, paper, photographic intermediates, plastics, polyurethanes, and textiles. It is frequently used to neutralize fatty acids and sulfonic acidbased surfactants.
Solubility
Soluble in water 20°C 870g/L. Miscible in alcohol.
Notes
Air Sensitive & Hygroscopic. Store away from oxidizing agents, air, water/moisture and light. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
Chemical Identifiers
110-97-4 | |
133.19 | |
LVTYICIALWPMFW-UHFFFAOYNA-N | |
8086 | |
CC(O)CNCC(C)O |
C6H15NO2 | |
MFCD00004531 | |
diisopropanolamine, 1,1'-iminodipropan-2-ol, bis 2-hydroxypropyl amine, 2-propanol, 1,1'-iminobis, bis 2-propanol amine, 1,1'-iminodi-2-propanol, dipa alcohol, di-2-propanolamine, 1,1'-azanediylbis propan-2-ol, dipropyl-2,2'-dihydroxy-amine | |
1-[(2-hydroxypropyl)amino]propan-2-ol |
Specifications
110-97-4 | |
1.004 | |
130°C (266°F) | |
C6H15NO2 | |
605363 | |
14,3195 | |
Soluble in water 20°C 870g/L. Miscible in alcohol. | |
CC(O)CNCC(C)O | |
133.19 | |
133.19 | |
Diisopropanolamine, sum of isomers |
38°C to 45°C | |
249°C to 250°C | |
Amine-like | |
MFCD00004531 | |
Air sensitive; Hygroscopic | |
diisopropanolamine, 1,1'-iminodipropan-2-ol, bis 2-hydroxypropyl amine, 2-propanol, 1,1'-iminobis, bis 2-propanol amine, 1,1'-iminodi-2-propanol, dipa alcohol, di-2-propanolamine, 1,1'-azanediylbis propan-2-ol, dipropyl-2,2'-dihydroxy-amine | |
LVTYICIALWPMFW-UHFFFAOYNA-N | |
1-[(2-hydroxypropyl)amino]propan-2-ol | |
8086 | |
≥98% |
Safety and Handling
GHS H Statement
H319
Causes serious eye irritation.
P264b-P280i-P305+P351+P338
H319
missing translation for 'einecsNumber' : 203-820-9
missing translation for 'rtecsNumber' : UB6600000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Argon
- Diisopropanolamine
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : Diisopropanolamine
RUO – Research Use Only