Learn More
Diisopropanolamine, 99%, Thermo Scientific Chemicals
£37.60 - £96.40
Chemical Identifiers
CAS | 110-97-4 |
---|---|
Molecular Formula | C6H15NO2 |
Molecular Weight (g/mol) | 133.19 |
MDL Number | MFCD00004531 |
InChI Key | LVTYICIALWPMFW-UHFFFAOYNA-N |
Synonym | diisopropanolamine, 1,1'-iminodipropan-2-ol, bis 2-hydroxypropyl amine, 2-propanol, 1,1'-iminobis, bis 2-propanol amine, 1,1'-iminodi-2-propanol, dipa alcohol, di-2-propanolamine, 1,1'-azanediylbis propan-2-ol, dipropyl-2,2'-dihydroxy-amine |
PubChem CID | 8086 |
IUPAC Name | 1-[(2-hydroxypropyl)amino]propan-2-ol |
SMILES | CC(O)CNCC(C)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10259640
|
Thermo Scientific Acros
407805000 |
500 g | Glass bottle |
£37.60
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10687274
|
Thermo Scientific Acros
407800025 |
2.5 kg | Glass bottle |
£96.40
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
110-97-4 | |
133.19 | |
LVTYICIALWPMFW-UHFFFAOYNA-N | |
8086 | |
CC(O)CNCC(C)O |
C6H15NO2 | |
MFCD00004531 | |
diisopropanolamine, 1,1'-iminodipropan-2-ol, bis 2-hydroxypropyl amine, 2-propanol, 1,1'-iminobis, bis 2-propanol amine, 1,1'-iminodi-2-propanol, dipa alcohol, di-2-propanolamine, 1,1'-azanediylbis propan-2-ol, dipropyl-2,2'-dihydroxy-amine | |
1-[(2-hydroxypropyl)amino]propan-2-ol |
Specifications
110-97-4 | |
Colorless to Yellow | |
249°C to 250°C | |
Authentic | |
C6H15NO2 | |
1.4615 to 1.4635 | |
15, 3214 | |
diisopropanolamine, 1,1'-iminodipropan-2-ol, bis 2-hydroxypropyl amine, 2-propanol, 1,1'-iminobis, bis 2-propanol amine, 1,1'-iminodi-2-propanol, dipa alcohol, di-2-propanolamine, 1,1'-azanediylbis propan-2-ol, dipropyl-2,2'-dihydroxy-amine | |
LVTYICIALWPMFW-UHFFFAOYNA-N | |
1-[(2-hydroxypropyl)amino]propan-2-ol | |
8086 | |
133.19 | |
Low Melting Solid |
33°C | |
1.0038g/mL | |
126°C | |
98.5 % min. (Total base) | |
(CH3CHOHCH2)2NH | |
MFCD00004531 | |
1.0038 | |
Solubility in water: 870g/L (20°C) | |
CC(O)CNCC(C)O | |
133.19 | |
84.19 mPa.s (60°C) | |
99% | |
Diisopropanolamine, 99% |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 203-820-9
missing translation for 'rtecsNumber' : UB6600000
missing translation for 'tsca' : TSCA
- Diisopropanolamine
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only