Learn More
Diisopropyl phosphite, 98%, Thermo Scientific Chemicals
£35.50 - £101.00
Chemical Identifiers
CAS | 1809-20-7 |
---|---|
Molecular Formula | C6H14O3P- |
Molecular Weight (g/mol) | 165.149 |
MDL Number | MFCD00117905 |
InChI Key | IWEDBEZKWHRUJX-UHFFFAOYSA-N |
Synonym | diisopropylphosphite, acmc-1bsaz, phosphorous acid diisopropyl anion, phosphorous acid, bis 1-methylethyl ester |
PubChem CID | 21884976 |
IUPAC Name | dipropan-2-yl phosphite |
SMILES | CC(C)OP([O-])OC(C)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11470258
|
Thermo Scientific Alfa Aesar
B23800.14 |
25 g |
£35.50
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11480258
|
Thermo Scientific Alfa Aesar
B23800.22 |
100 g |
£101.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Diisopropyl phosphite is used as antiwear lubricant additive together with triphenyl thiophosphate on T8 steel/Al2O3 ceramics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDiisopropyl phosphite is used as antiwear lubricant additive together with triphenyl thiophosphate on T8 steel/Al2O3 ceramics.
Solubility
Soluble in water, chloroform.
Notes
Moisture sensitive. Store under inert gas. Store away from oxidizing agents, water/ moisture.
Chemical Identifiers
1809-20-7 | |
165.149 | |
IWEDBEZKWHRUJX-UHFFFAOYSA-N | |
21884976 | |
CC(C)OP([O-])OC(C)C |
C6H14O3P- | |
MFCD00117905 | |
diisopropylphosphite, acmc-1bsaz, phosphorous acid diisopropyl anion, phosphorous acid, bis 1-methylethyl ester | |
dipropan-2-yl phosphite |
Specifications
1809-20-7 | |
72°C to 75°C (10 mmHg) | |
C6H14O3P- | |
1.4075 | |
Moisture sensitive | |
Soluble in water,chloroform. | |
CC(C)OP([O-])OC(C)C | |
165.149 | |
166.16 | |
Diisopropyl phosphite |
0.99 | |
112°C (234°F) | |
[(CH3)2CHO]2P(O)H | |
MFCD00117905 | |
diisopropylphosphite, acmc-1bsaz, phosphorous acid diisopropyl anion, phosphorous acid, bis 1-methylethyl ester | |
IWEDBEZKWHRUJX-UHFFFAOYSA-N | |
dipropan-2-yl phosphite | |
21884976 | |
98% |
Safety and Handling
GHS H Statement
H301-H332-H315-H319-H335
Toxic if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'einecsNumber' : 217-317-7
missing translation for 'rtecsNumber' : SZ7660000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only