missing translation for 'onlineSavingsMsg'
Learn More

Diisopropyl sulfide, 99%, Thermo Scientific Chemicals

Product Code. 11409217
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Used as a flavoring agents and intermediate for organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as a flavoring agents and intermediate for organic synthesis.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 625-80-9
Molecular Formula C6H14S
Molecular Weight (g/mol) 118.24
MDL Number MFCD00008895
InChI Key XYWDPYKBIRQXQS-UHFFFAOYSA-N
Synonym diisopropyl sulfide, isopropyl sulfide, propane, 2,2'-thiobis, 2,4-dimethyl-3-thiapentane, isopropyl sulphide, diisopropyl sulphide, 2,2'-thiodipropane, 2-isopropylsulfanyl propane, 2-propan-2-ylsulfanyl propane, isopropylsulfide
PubChem CID 12264
SMILES CC(C)SC(C)C

Specifications

Density 0.814
Boiling Point 119°C to 120°C
Flash Point 7°C (44°F)
Odor Stench
Linear Formula (CH3)2CHSCH(CH3)2
Refractive Index 1.4385
Quantity 50 g
UN Number UN1993
Beilstein 1731257
Solubility Information Not miscible or difficult to mix with water.
IUPAC Name 2-(propan-2-ylsulfanyl)propane
Formula Weight 118.24
Percent Purity 99%
Chemical Name or Material Diisopropyl sulfide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'