Learn More
Diisopropylamine, 99.5%, redistilled, AcroSeal™, Thermo Scientific Chemicals
£55.98 - £270.35
Chemical Identifiers
CAS | 108-18-9 |
---|---|
Molecular Formula | C6H15N |
Molecular Weight (g/mol) | 101.19 |
MDL Number | MFCD00008862 |
InChI Key | UAOMVDZJSHZZME-UHFFFAOYSA-N |
Synonym | diisopropylamine, 2-propanamine, n-1-methylethyl, n-1-methylethyl-2-propanamine, dipa, n,n-diisopropylamine, bis propan-2-yl amine, n-isopropylpropan-2-amine, di-isopropylamine, n-isopropyl-1-amino-2-methylethane, diisopropyl amine |
PubChem CID | 7912 |
IUPAC Name | N-propan-2-ylpropan-2-amine |
SMILES | CC(C)NC(C)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10308472
|
Thermo Scientific Acros
378301000 |
100 mL | AcroSeal™ Glass Bottle |
£55.98
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10766573
|
Thermo Scientific Acros
378300010 |
1 L | AcroSeal™ Glass Bottle |
£270.35
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
108-18-9 | |
101.19 | |
UAOMVDZJSHZZME-UHFFFAOYSA-N | |
7912 | |
CC(C)NC(C)C |
C6H15N | |
MFCD00008862 | |
diisopropylamine, 2-propanamine, n-1-methylethyl, n-1-methylethyl-2-propanamine, dipa, n,n-diisopropylamine, bis propan-2-yl amine, n-isopropylpropan-2-amine, di-isopropylamine, n-isopropyl-1-amino-2-methylethane, diisopropyl amine | |
N-propan-2-ylpropan-2-amine |
Specifications
108-18-9 | |
0.7160g/mL | |
−17°C | |
99.4% min. (GC) | |
[(CH3)2CH]2NH | |
MFCD00008862 | |
0.716 | |
Solubility in water: 100g/L (20°C). Other solubilities: soluble in alcohol,soluble in most common organic solvents | |
CC(C)NC(C)C | |
101.19 | |
0.4 mPa.s (20°C) | |
99.5% | |
Diisopropylamine |
-61.0°C | |
84.0°C | |
Authentic | |
C6H15N | |
1.3910 to 1.3930 | |
15, 3215 | |
diisopropylamine, 2-propanamine, n-1-methylethyl, n-1-methylethyl-2-propanamine, dipa, n,n-diisopropylamine, bis propan-2-yl amine, n-isopropylpropan-2-amine, di-isopropylamine, n-isopropyl-1-amino-2-methylethane, diisopropyl amine | |
UAOMVDZJSHZZME-UHFFFAOYSA-N | |
N-propan-2-ylpropan-2-amine | |
7912 | |
101.19 | |
Liquid |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Toxic if inhaled.
May cause respiratory irritation.
Highly flammable liquid and vapor.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 203-558-5
- Diisopropylamine
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Acute toxicity Category 4
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H331-Toxic if inhaled.
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only