Learn More
Dilauroyl peroxide, 99%, Thermo Scientific Chemicals
£27.60 - £86.10
Chemical Identifiers
CAS | 105-74-8 |
---|---|
Molecular Formula | C24H46O4 |
Molecular Weight (g/mol) | 398.63 |
MDL Number | MFCD00008964 |
InChI Key | YIVJZNGAASQVEM-UHFFFAOYSA-N |
Synonym | lauroyl peroxide, dilauroyl peroxide, dodecanoyl peroxide, peroxide, bis 1-oxododecyl, laurydol, laurox, alperox c, dodecanoic peroxyanhydride, dyp-97f, peroxyde de lauroyle |
PubChem CID | 7773 |
ChEBI | CHEBI:82479 |
IUPAC Name | dodecanoyl dodecaneperoxoate |
SMILES | CCCCCCCCCCCC(=O)OOC(=O)CCCCCCCCCCC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10377021
|
Thermo Scientific Acros
349741000 |
100 g | Plastic bottle |
£27.60
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10643512
|
Thermo Scientific
349745000 |
500 g | Plastic bottle |
£86.10
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Radical initiatorsChemical Identifiers
105-74-8 | |
398.63 | |
YIVJZNGAASQVEM-UHFFFAOYSA-N | |
7773 | |
dodecanoyl dodecaneperoxoate |
C24H46O4 | |
MFCD00008964 | |
lauroyl peroxide, dilauroyl peroxide, dodecanoyl peroxide, peroxide, bis 1-oxododecyl, laurydol, laurox, alperox c, dodecanoic peroxyanhydride, dyp-97f, peroxyde de lauroyle | |
CHEBI:82479 | |
CCCCCCCCCCCC(=O)OOC(=O)CCCCCCCCCCC |
Specifications
105-74-8 | |
White | |
98.5% min. (Iodometry) | |
[CH3(CH2)10CO]2O2 | |
lauroyl peroxide, dilauroyl peroxide, dodecanoyl peroxide, peroxide, bis 1-oxododecyl, laurydol, laurox, alperox c, dodecanoic peroxyanhydride, dyp-97f, peroxyde de lauroyle | |
YIVJZNGAASQVEM-UHFFFAOYSA-N | |
dodecanoyl dodecaneperoxoate | |
7773 | |
398.63 | |
Powder |
53°C to 57°C | |
Authentic | |
C24H46O4 | |
MFCD00008964 | |
Solubility in water: insoluble. Other solubilities: partly soluble in toluene and hexane | |
CCCCCCCCCCCC(=O)OOC(=O)CCCCCCCCCCC | |
398.63 | |
CHEBI:82479 | |
99% | |
Dilauroyl peroxide, 99% |
Safety and Handling
GHS H Statement
Heating may cause a fire.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Keep/Store away from clothing/ combustible materials.
Keep only in orig
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 203-326-3
missing translation for 'rtecsNumber' : OF2625000
missing translation for 'tsca' : TSCA
- Dilauroyl peroxide
Signal Word
- Danger
Hazard Category
- Organic peroxide Type C & D
Hazard Statement
- H242-Heating may cause a fire.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P220-Keep/Store away from clothing/combustible materials.
- P234-Keep only in original container.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P403+P235-Store in a well-ventilated place. Keep cool.
RUO – Research Use Only