missing translation for 'onlineSavingsMsg'
Learn More

Dilithium tetrabromonickelate(II), 0.5M solution in THF, AcroSeal™, Thermo Scientific Chemicals

Product Code. 11954341
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13826-95-4, 109-99-9
Molecular Formula Br4Li2Ni
Molecular Weight (g/mol) 392.19
InChI Key AVJDNPHXGKKFIA-UHFFFAOYSA-J
Synonym di-lithium tetrabromonickelate ii, agn-pc-0nf0dx, lithium tetrabromonickelate ii solution, dilithium tetrabromonickelate ii solution
PubChem CID 11047451
IUPAC Name dilithium;tetrabromonickel(2-)
SMILES [Li+].[Li+].[Ni-2](Br)(Br)(Br)Br

Specifications

Color Blue-Green to Brown
Physical Form Solution
Quantity 100 mL
Linear Formula Li2NiBr4
Flash Point −18°C
Formula Weight 392.19
Packaging AcroSeal™ Glass Bottle
Specific Gravity 1.065
Density 1.0650g/mL
Chemical Name or Material Dilithium tetrabromonickelate(II)
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Dilithium tetrabromonickelate(II), 0.5M solution in THF
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H350-May cause cancer.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.