missing translation for 'onlineSavingsMsg'
Learn More

Dilithium tetrachlorocuprate, 0.1M solution in THF, Thermo Scientific Chemicals

£24.12 - £197.99

Specifications

Chemical Name or Material Dilithium tetrachlorocuprate
Color Colorless
Name Note 0.1M Solution in THF
CAS 15489-27-7, 109-99-9
Molecular Formula Cl4CuLi2
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10112241
View Documents Promotion Details
Thermo Scientific Acros
407830250
25 mL Glass bottle
£24.12
25mL
Please to purchase this item. Need a web account? Register with us today!
10346401
View Documents
Thermo Scientific Acros
407831000
100 mL Glass bottle
£61.81
100mL
Please to purchase this item. Need a web account? Register with us today!
10420831
View Documents
Thermo Scientific Acros
407835000
500 mL Glass bottle
£197.99
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Dilithium tetrachlorocuprate
0.1M Solution in THF
Cl4CuLi2
MFCD00011081
04,163; 05,226; 06,203; 07,114; 08,176; 11,190; 12,195
HCJWWBBBSCXJMS-UHFFFAOYSA-J
dilithium;tetrachlorocopper(2-)
11074879
Glass bottle
−17°C
Colorless
15489-27-7, 109-99-9
Li2CuCl4
0.09 to 0.12M
0.9100g/mL
[Li+].[Li+].Cl[Cu-2](Cl)(Cl)Cl
219.226
219.24
0.91
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation. Causes serious eye irritation. Highly flammable liquid and vapor. May form explosive peroxides. Suspected of causing cancer. Harmful if swallowed. May cause drowsiness or dizziness.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Call a POISON CENTER or doctor/physician if you feel unwell. Wash face, hands and any exposed skin thoroughly after handling. If eye irritation persists: Get medical advice/attention. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Wear protective gloves/protective clothing/eye protection/face protection.

Danger

Product Identifier
  • Dilithium tetrachlorocuprate, 0.1M solution in THF

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
  • EUH019-May form explosive peroxides.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.