missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Diltiazem hydrochloride, 98% Product Code.: 10276161

Thermo Scientific Chemicals Diltiazem hydrochloride, 98%

Product Code. 10276161
1 g, Glass bottle
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10276161

Brand: Thermo Scientific Chemicals 329080010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33286-22-5
Molecular Formula C22H26N2O4S·HCl
Molecular Weight (g/mol) 450.99
MDL Number MFCD00069252
InChI Key HDRXZJPWHTXQRI-BHDTVMLSSA-N
Synonym diltiazem hydrochloride, diltiazem hcl, herbesser, dilzene, lacerol, masdil, tildiem, tiazac, mono-tildiem, bi-tildiem
PubChem CID 62920
ChEBI CHEBI:645509
IUPAC Name [(2S,3S)-5-[2-(dimethylamino)ethyl]-2-(4-methoxyphenyl)-4-oxo-2,3-dihydro-1,5-benzothiazepin-3-yl] acetate;hydrochloride
SMILES CC(=O)OC1C(SC2=CC=CC=C2N(C1=O)CCN(C)C)C3=CC=C(C=C3)OC.Cl

Specifications

Melting Point 210.0°C to 215.0°C
Color White
Ignition Residue 0.1% max.
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Merck Index 15,3224
Solubility Information Solubility in water: soluble. Other solubilities: soluble in chloroform,formic acid,methanol
Formula Weight 450.99
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Diltiazem hydrochloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0463761 Certificate TypeCertificate of Analysis Date18/06/2024 Catalog Number329080010,329080050,329080250
Lot NumberA0403611 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number329080010,329080050,329080250
Lot NumberA0447909 Certificate TypeCertificate of Analysis Date09/12/2022 Catalog Number329080010,329080050,329080250
Lot NumberA0414306 Certificate TypeCertificate of Analysis Date30/05/2022 Catalog Number329080010,329080050,329080250
Lot NumberA0431457 Certificate TypeCertificate of Analysis Date22/06/2021 Catalog Number329080010,329080050,329080250
compliance-icons
Product Identifier
  • Dilthiazem hydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Diltiazem hydrochloride, 98% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.