missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Antihypertensive and cardioprotective agent; an inhibitor of L-type Ca2+ channels.

Specifications
Specifications
CAS | 33286-22-5 |
Molecular Formula | C22H27ClN2O4S |
Quantity | 1 g |
Synonym | diltiazem hydrochloride, diltiazem hcl, herbesser, dilzene, lacerol, masdil, tildiem, tiazac, mono-tildiem, bi-tildiem |
InChI Key | HDRXZJPWHTXQRI-BHDTVMLSSA-N |
SMILES | CC(=O)OC1C(SC2=CC=CC=C2N(C1=O)CCN(C)C)C3=CC=C(C=C3)OC.Cl |
IUPAC Name | [(2S,3S)-5-[2-(dimethylamino)ethyl]-2-(4-methoxyphenyl)-4-oxo-2,3-dihydro-1,5-benzothiazepin-3-yl] acetate;hydrochloride |
Molecular Weight (g/mol) | 450.978 |
PubChem CID | 62920 |
ChEBI | CHEBI:645509 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Diltiazem hydrochloride, Tocris Bioscience™ >
Spot an opportunity for improvement?Share a Content Correction