Learn More
Implen™ DiluCell™ 10 - Disposable Cuvette
Implen DiluCell™ 10 provides an automatic 1/10 dilution of higher concentrated samples for standard 1 cm cuvette spectrophotometers. Combined with low sample volume requirements and bubble free filling, the DiluCell™ allows for convenient spectrophotometric analysis from 340-1100 nm.
Brand:  Implen™ DC 10
Product Code. 15725805
  • £101.21 / Each
Estimated Shipment: 04-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Implen DiluCell™… make it virtually dilute your sample!
Previously it was necessary to use standard cuvettes with a volume of at least 1 mL and additionally to dilute the sample manually. DiluCell™ from Implen allows for lower sample volume requirements and provides automatic virtual sample dilution. Due to the unique design of DiluCell™ cuvettes, the light-path is reduced from the standard 10 mm to 0.5 mm (DC 20) or 1.0 mm (DC 10). According to the Lambert Beer Law this shortened pathlength results in an automatic sample dilution by factor 20 (DC 20) or 10 (DC 10). Automatic dilution with DiluCell™ saves time and excludes dilution errors and cross contaminations.
  • Low sample volume necessary
  • Suitable for proteins and microbial cell cultures
  • Measurement possible starting at 340 nm
  • Automatic dilution of factor 10 and factor 20
  • No risk of contamination
  • Significantly delayed cell sedimentation compared to standard cuvettes
  • Approved for usage with the Implen OD600 DiluPhotometer™
  • Made in Germany
Specifications
Specifications
1/10 Dilution
200 μL
Pack of 96
340 to 1100 nm
Protein assays, OD600
Single-use
Plastic
1.0 mm
Spectrophotometer, OD600 DiluPhotometer
12 x 12 x 45 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Implen™ DiluCell™ 10 - Disposable Cuvette > Capacity (Metric): 200 μL