missing translation for 'onlineSavingsMsg'
Learn More

Dimedone, 98%, Thermo Scientific Chemicals

£20.60 - £199.00

Chemical Identifiers

CAS 126-81-8
Molecular Formula C8H12O2
Molecular Weight (g/mol) 140.18
MDL Number MFCD00001588
InChI Key BADXJIPKFRBFOT-UHFFFAOYSA-N
Synonym dimedone, 5,5-dimethyl-1,3-cyclohexanedione, cyclomethone, medon, methone, dimedon, methon, 1,3-cyclohexanedione, 5,5-dimethyl, 5,5-dimethyldihydroresorcinol, 5,5-dimethylhydroresorcinol
PubChem CID 31358
IUPAC Name 5,5-dimethylcyclohexane-1,3-dione
SMILES CC1(C)CC(=O)CC(=O)C1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11469402
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A10140.14
25 g
£20.60
25g
Please to purchase this item. Need a web account? Register with us today!
11479402
View Documents
Thermo Scientific Alfa Aesar
A10140.22
100 g
£55.68
100g
Please to purchase this item. Need a web account? Register with us today!
11489402
View Documents
Thermo Scientific Alfa Aesar
A10140.36
500 g
£199.00
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Reagent used in synthesis of fused 3-methylfuransDimedone is used in organic chemistry to identify the presence of aldehyde group. It is also used as a catalyst in the formation of transition-metal complexes. Further, it is also employed in colorimetry, crystallography, luminescence and spectrophotometric analysis. In addition to this, it is used in the synthesis of naturally occurring fused 3-methylfurans.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reagent used in synthesis of fused 3-methylfuransDimedone is used in organic chemistry to identify the presence of aldehyde group. It is also used as a catalyst in the formation of transition-metal complexes. Further, it is also employed in colorimetry, crystallography, luminescence and spectrophotometric analysis. In addition to this, it is used in the synthesis of naturally occurring fused 3-methylfurans.

Solubility
Soluble in methanol, ethanol, benzene and chloroform. Slightly soluble in water.

Notes
Light sensitive. Incompatible with oxidizing agents, strong acids and strong bases.
Specifications

Chemical Identifiers

126-81-8
140.18
BADXJIPKFRBFOT-UHFFFAOYSA-N
31358
CC1(C)CC(=O)CC(=O)C1
C8H12O2
MFCD00001588
dimedone, 5,5-dimethyl-1,3-cyclohexanedione, cyclomethone, medon, methone, dimedon, methon, 1,3-cyclohexanedione, 5,5-dimethyl, 5,5-dimethyldihydroresorcinol, 5,5-dimethylhydroresorcinol
5,5-dimethylcyclohexane-1,3-dione

Specifications

126-81-8
C8H12O2
471489
dimedone, 5,5-dimethyl-1,3-cyclohexanedione, cyclomethone, medon, methone, dimedon, methon, 1,3-cyclohexanedione, 5,5-dimethyl, 5,5-dimethyldihydroresorcinol, 5,5-dimethylhydroresorcinol
BADXJIPKFRBFOT-UHFFFAOYSA-N
5,5-dimethylcyclohexane-1,3-dione
31358
98%
146°C to 150°C
MFCD00001588
14,3242
Soluble in methanol,ethanol,benzene and chloroform. Slightly soluble in water.
CC1(C)CC(=O)CC(=O)C1
140.18
140.18
Dimedone
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 204-804-4

missing translation for 'rtecsNumber' : GV0345000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only