missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethoxymethane, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Description
Used to protect diols as their methylene acetals, using TMS-OTf/2,6-lutidine:
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Used to protect diols as their methylene acetals, using TMS-OTf/2,6-lutidine:
Solubility
Soluble in water 20°C 330g/L.
Notes
Moisture Sensitive. Store away from oxidizing agents, moisture/water and acids. Store under inert gas.
Used to protect diols as their methylene acetals, using TMS-OTf/2,6-lutidine:
Solubility
Soluble in water 20°C 330g/L.
Notes
Moisture Sensitive. Store away from oxidizing agents, moisture/water and acids. Store under inert gas.
Specifications
Specifications
Melting Point | -105°C |
Density | 0.863 |
Boiling Point | 41°C to 42°C |
Flash Point | −17°C (1°F) |
Linear Formula | CH3OCH2OCH3 |
Refractive Index | 1.3545 |
Quantity | 100 mL |
UN Number | UN1234 |
Beilstein | 1697025 |
Sensitivity | Moisture sensitive |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'