missing translation for 'onlineSavingsMsg'
Learn More

Dimethoxymethane, 99.5+%, Thermo Scientific Chemicals

Product Code. 10073563
Click to view available options
Quantity:
1 L
2.5 L
25 L
Packaging:
Glass bottle
Metal drum
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-87-5
Molecular Formula C3H8O2
Molecular Weight (g/mol) 76.09
MDL Number MFCD00008495
InChI Key NKDDWNXOKDWJAK-UHFFFAOYSA-N
Synonym methylal, formal, methane, dimethoxy, formaldehyde dimethyl acetal, dimethyl formal, anesthenyl, 2,4-dioxapentane, bis methoxy methane, methylene dimethyl ether, methoxymethyl methyl ether
PubChem CID 8020
ChEBI CHEBI:48341
IUPAC Name dimethoxymethane
SMILES COCOC

Specifications

Melting Point -105.0°C
Color Colorless
Density 0.8600g/mL
Boiling Point 41.0°C to 42.0°C
Flash Point −18°C
Infrared Spectrum Authentic
Assay Percent Range 99.5% min. (GC)
Packaging Metal drum
Linear Formula CH2(OCH3)2
Refractive Index 1.3525 to 1.3545
Quantity 25 L
Fieser 01,671; 02,271; 06,374; 07,115
Merck Index 15, 6086
Specific Gravity 0.86
Solubility Information Solubility in water: 32.3g/100 mL (16°C). Other solubilities: miscible with alcohol,ether,oils,hydrocarbons,miscible with most common organic solvents,very soluble in acetone,benzene
Viscosity 3.25 mPa.s (20°C)
Formula Weight 76.09
Percent Purity 99.5+%
Physical Form Liquid
Chemical Name or Material Dimethoxymethane
Show More Show Less
compliance-icons
Product Identifier
  • Dimethoxymethane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.