Learn More
Dimethoxymethyl(3,3,3-trifluoropropyl)silane, 97%, Thermo Scientific Chemicals
£43.90 - £130.00
Chemical Identifiers
CAS | 358-67-8 |
---|---|
Molecular Formula | C6H13F3O2Si |
Molecular Weight (g/mol) | 202.248 |
MDL Number | MFCD00053175 |
InChI Key | DIJRHOZMLZRNLM-UHFFFAOYSA-N |
Synonym | dimethoxy methyl 3,3,3-trifluoropropyl silane, 3,3,3-trifluoropropyl methyldimethoxysilane, 3,3,3-trifluoropropylmethyldimethoxysilane, dimethoxymethyl 3,3,3-trifluoropropyl silane, silane, dimethoxymethyl 3,3,3-trifluoropropyl, dimethoxy-methyl-3,3,3-trifluoropropyl silane, acmc-1cm60, 3,3,3-trifluoropropyldimethoxy methyl silane, methyl 3,3,3-trifluoropropyl dimethoxysilane, silane,dimethoxymethyl 3,3,3-trifluoropropyl |
PubChem CID | 67751 |
IUPAC Name | dimethoxy-methyl-(3,3,3-trifluoropropyl)silane |
SMILES | CO[Si](C)(CCC(F)(F)F)OC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11344026
|
Thermo Scientific Alfa Aesar
L17292.06 |
5 g |
£43.90
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11358376
|
Thermo Scientific Alfa Aesar
L17292.14 |
25 g |
£130.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
358-67-8 | |
202.248 | |
DIJRHOZMLZRNLM-UHFFFAOYSA-N | |
67751 | |
CO[Si](C)(CCC(F)(F)F)OC |
C6H13F3O2Si | |
MFCD00053175 | |
dimethoxy methyl 3,3,3-trifluoropropyl silane, 3,3,3-trifluoropropyl methyldimethoxysilane, 3,3,3-trifluoropropylmethyldimethoxysilane, dimethoxymethyl 3,3,3-trifluoropropyl silane, silane, dimethoxymethyl 3,3,3-trifluoropropyl, dimethoxy-methyl-3,3,3-trifluoropropyl silane, acmc-1cm60, 3,3,3-trifluoropropyldimethoxy methyl silane, methyl 3,3,3-trifluoropropyl dimethoxysilane, silane,dimethoxymethyl 3,3,3-trifluoropropyl | |
dimethoxy-methyl-(3,3,3-trifluoropropyl)silane |
Specifications
85°C (150mmHg) | |
C6H13F3O2Si | |
UN1993 | |
58°C (136°F) | |
DIJRHOZMLZRNLM-UHFFFAOYSA-N | |
dimethoxy-methyl-(3,3,3-trifluoropropyl)silane | |
67751 | |
97% | |
Moisture sensitive | |
Dimethoxymethyl(3,3,3-trifluoropropyl)silane |
358-67-8 | |
MFCD00053175 | |
1753237 | |
dimethoxy methyl 3,3,3-trifluoropropyl silane, 3,3,3-trifluoropropyl methyldimethoxysilane, 3,3,3-trifluoropropylmethyldimethoxysilane, dimethoxymethyl 3,3,3-trifluoropropyl silane, silane, dimethoxymethyl 3,3,3-trifluoropropyl, dimethoxy-methyl-3,3,3-trifluoropropyl silane, acmc-1cm60, 3,3,3-trifluoropropyldimethoxy methyl silane, methyl 3,3,3-trifluoropropyl dimethoxysilane, silane,dimethoxymethyl 3,3,3-trifluoropropyl | |
CO[Si](C)(CCC(F)(F)F)OC | |
202.248 | |
202.25 | |
1.3576 | |
1.1 |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
missing translation for 'einecsNumber' : 206-619-4
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only