missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl 1,4-cyclohexanedione-2,5-dicarboxylate, 99+%, Thermo Scientific Chemicals

Catalog Number 10553411
Click to view available options
Quantity:
50 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
6289-46-9
C10H12O6
228.20
MFCD00001607
MHKKFFHWMKEBDW-UHFFFAOYNA-N
dimethyl succinylsuccinate, dimethyl 1,4-cyclohexanedione-2,5-dicarboxylate, succinosuccinic acid, dimethyl ester, dmss, dimethyl succinylosuccinate, dimethyl 2,5-dioxo-1,4-cyclohexanedicarboxylate, dimethyl succinylo succinate, 1,4-cyclohexanedicarboxylic acid, 2,5-dioxo-, dimethyl ester, 2,5-dioxo-1,4-cyclohexanedicarboxylic acid dimethyl ester, 1,4-cyclohexanedicarboxylic acid, 2,5-dioxo-, 1,4-dimethyl ester
94866
dimethyl 2,5-dioxocyclohexane-1,4-dicarboxylate
COC(=O)C1CC(=O)C(CC1=O)C(=O)OC
This item is not returnable. View return policy
6289-46-9
C10H12O6
228.20
MFCD00001607
MHKKFFHWMKEBDW-UHFFFAOYNA-N
dimethyl succinylsuccinate, dimethyl 1,4-cyclohexanedione-2,5-dicarboxylate, succinosuccinic acid, dimethyl ester, dmss, dimethyl succinylosuccinate, dimethyl 2,5-dioxo-1,4-cyclohexanedicarboxylate, dimethyl succinylo succinate, 1,4-cyclohexanedicarboxylic acid, 2,5-dioxo-, dimethyl ester, 2,5-dioxo-1,4-cyclohexanedicarboxylic acid dimethyl ester, 1,4-cyclohexanedicarboxylic acid, 2,5-dioxo-, 1,4-dimethyl ester
94866
dimethyl 2,5-dioxocyclohexane-1,4-dicarboxylate
COC(=O)C1CC(=O)C(CC1=O)C(=O)OC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 154°C to 157°C
Color Green-Yellow to White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Quantity 100 g
Beilstein 10, IV, 3514
Formula Weight 228.2
Percent Purity 99+%
Physical Form Fine Crystalline Powder
Chemical Name or Material Dimethyl 1, 4-cyclohexanedione-2, 5-dicarboxylate, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • Dimethyl 1,4-cyclohexanedione-2,5-dicarboxylate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'