missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(+)-Dimethyl 2,3-O-isopropylidene-D-tartrate, 97%, Thermo Scientific Chemicals
£96.90 - £448.00
Chemical Identifiers
CAS | 37031-30-4 |
---|---|
Molecular Formula | C9H14O6 |
Molecular Weight (g/mol) | 218.21 |
MDL Number | MFCD00040504 |
InChI Key | ROZOUYVVWUTPNG-WDSKDSINSA-N |
Synonym | 4s,5s-2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylic acid dimethyl ester, +-dimethyl 2,3-o-isopropylidene-d-tartrate, dimethyl 4s,5s-2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate, 4s,5s-dimethyl 2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate, 4,5-dimethyl 4s,5s-2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate, pubchem6765, dimethyl--2,3-o-isopropylidene-l-tartrate, dimethyl 2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate #, +-dimethyl-2,3-o-isopropylidine-d-tartrate, dimethyl +-2,3-o-isopropylidene-d-tartrate |
PubChem CID | 688156 |
IUPAC Name | dimethyl (4S,5S)-2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate |
SMILES | CC1(OC(C(O1)C(=O)OC)C(=O)OC)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10677611
|
Thermo Scientific Acros
258440100 |
10 mL | Glass bottle |
£96.90
10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10185833
|
Thermo Scientific Acros
258440500 |
50 mL | Glass bottle |
£448.00
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
37031-30-4 | |
218.21 | |
ROZOUYVVWUTPNG-WDSKDSINSA-N | |
688156 | |
CC1(OC(C(O1)C(=O)OC)C(=O)OC)C |
C9H14O6 | |
MFCD00040504 | |
4s,5s-2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylic acid dimethyl ester, +-dimethyl 2,3-o-isopropylidene-d-tartrate, dimethyl 4s,5s-2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate, 4s,5s-dimethyl 2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate, 4,5-dimethyl 4s,5s-2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate, pubchem6765, dimethyl--2,3-o-isopropylidene-l-tartrate, dimethyl 2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate #, +-dimethyl-2,3-o-isopropylidine-d-tartrate, dimethyl +-2,3-o-isopropylidene-d-tartrate | |
dimethyl (4S,5S)-2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate |
Specifications
37031-30-4 | |
100.0 | |
1.1500g/mL | |
135°C | |
96% min. (GC) | |
1.4386 to 1.4406 | |
1.15 | |
+ 54 | |
ROZOUYVVWUTPNG-WDSKDSINSA-N | |
dimethyl (4S,5S)-2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate | |
688156 | |
97% | |
(+)-Dimethyl 2, 3-O-isopropylidene-D-tartrate |
96.0 | |
Colorless to Yellow | |
139°C (21.0 hPa) | |
Authentic | |
C9H14O6 | |
MFCD00040504 | |
+ 54.00 (20.00°C c=neat) | |
4s,5s-2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylic acid dimethyl ester, +-dimethyl 2,3-o-isopropylidene-d-tartrate, dimethyl 4s,5s-2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate, 4s,5s-dimethyl 2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate, 4,5-dimethyl 4s,5s-2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate, pubchem6765, dimethyl--2,3-o-isopropylidene-l-tartrate, dimethyl 2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate #, +-dimethyl-2,3-o-isopropylidine-d-tartrate, dimethyl +-2,3-o-isopropylidene-d-tartrate | |
CC1(OC(C(O1)C(=O)OC)C(=O)OC)C | |
218.21 | |
218.21 | |
Liquid |
Safety and Handling
Product Identifier
- (+)-Dimethyl-2,3-O-isopropylidene-D-tartrate
RUO – Research Use Only