missing translation for 'onlineSavingsMsg'
Learn More
Dimethyl aminoterephthalate, 99%, Thermo Scientific Chemicals Product Code.: 11483214

Dimethyl aminoterephthalate, 99%, Thermo Scientific Chemicals

Product Code. 11483214
50 g
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy

Product Code. 11483214

Brand: Thermo Scientific Alfa Aesar A10567.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Dimethyl aminoterephthalate is used in the preparation of imidazotetrazinones as probes for the action of temozolomide. It is also used to prepare aminoquinazolines with neurokinin-2 receptor antagonist activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl aminoterephthalate is used in the preparation of imidazotetrazinones as probes for the action of temozolomide. It is also used to prepare aminoquinazolines with neurokinin-2 receptor antagonist activity.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5372-81-6
Molecular Formula C10H11NO4
Molecular Weight (g/mol) 209.201
MDL Number MFCD00008427
InChI Key DSSKDXUDARIMTR-UHFFFAOYSA-N
Synonym dimethyl 2-aminoterephthalate, dimethyl aminoterephthalate, dimethyl 3-aminoterephthalate, 1,4-benzenedicarboxylic acid, 2-amino-, dimethyl ester, aminoterephthalic acid dimethyl ester, 1,4-dimethyl 2-aminobenzene-1,4-dicarboxylate, unii-91sf4e6i9w, 2-aminoterephthalic acid, dimethyl ester, 3-amino-4-methoxycarbonylbenzoic acid, methyl ester, terephthalic acid, amino-, dimethyl ester
PubChem CID 79336
IUPAC Name dimethyl 2-aminobenzene-1,4-dicarboxylate
SMILES COC(=O)C1=CC(=C(C=C1)C(=O)OC)N

Specifications

Melting Point 130°C to 134°C
Quantity 50 g
Beilstein 2808505
Solubility Information Insoluble in water.
Formula Weight 209.2
Percent Purity 99%
Chemical Name or Material Dimethyl aminoterephthalate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dimethyl aminoterephthalate, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.