missing translation for 'onlineSavingsMsg'
Learn More
Dimethyl carbonate, 99+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals Product Code.: 13198473

Dimethyl carbonate, 99+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals

Product Code. 13198473
100 mL, AcroSeal™ Glass Bottle
Click to view available options
Quantity:
100 mL
1 L
Packaging:
AcroSeal™ Glass Bottle
This item is not returnable. View return policy

Product Code. 13198473

Brand: Thermo Scientific Acros 428731000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 616-38-6
Molecular Formula C3H6O3
Molecular Weight (g/mol) 90.08
MDL Number MFCD00008420
InChI Key IEJIGPNLZYLLBP-UHFFFAOYSA-N
Synonym methyl carbonate, carbonic acid, dimethyl ester, carbonic acid dimethyl ester, methyl carbonate meo 2co, unii-ke9j097spn, dimethylcarbonate, ch3ocooch3, ke9j097spn, dimethyl ester of carbonic acid, dsstox_cid_9192Show MoreShow Less
PubChem CID 12021
ChEBI CHEBI:36596
IUPAC Name dimethyl carbonate
SMILES COC(=O)OC

Specifications

Melting Point 2.0°C to 4.0°C
Color Brown to White
Density 1.0690g/mL
Boiling Point 90.0°C
Flash Point 18°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging AcroSeal™ Glass Bottle
Linear Formula (CH3O)2CO
Refractive Index 1.3672 to 1.3692
Quantity 100 mL
Merck Index 15, 3263
Specific Gravity 1.069
Solubility Information Solubility in water: 139g/L. Other solubilities: soluble in all proportions with organic alcohols,soluble in all proportions with esters and ketones,miscible with acids and alkalies
Viscosity 0.625 mPa.s (20°C)
Formula Weight 90.08
Percent Purity 99+%
Grade Extra Dry
Physical Form Crystalline Powder
Chemical Name or Material Dimethyl carbonate, Extra Dry
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2400806 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number428731000,428730010
Lot Number2498632 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number428731000,428730010
Lot Number2524013 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number428731000,428730010
Lot Number2400805 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number428731000,428730010
Lot Number2524012 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number428731000,428730010
compliance-icons
Product Identifier
  • Dimethyl carbonate
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dimethyl carbonate, 99+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.