missing translation for 'onlineSavingsMsg'
Learn More
Dimethyl cis-1,2-cyclopropanedicarboxylate, 97+%, Thermo Scientific Chemicals Product Code.: 10195593

Dimethyl cis-1,2-cyclopropanedicarboxylate, 97+%, Thermo Scientific Chemicals

Product Code. 10195593
25 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10195593

Brand: Thermo Scientific Acros 255340250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 826-34-6
Molecular Formula C7H10O4
Molecular Weight (g/mol) 158.16
MDL Number MFCD00062805
InChI Key JBVOSZYUSFDYIN-SYDPRGILSA-N
Synonym dimethyl cis-1,2-cyclopropanedicarboxylate, dimethyl 1r,2s-cyclopropane-1,2-dicarboxylate, cis-dimethyl cyclopropane-1,2-dicarboxylate, dimethylcis-1,2-cyclopropanedicarboxylate, cyclopropane-1,2-dicarboxylic acid dimethyl ester, z, 1,2-cyclopropanedicarboxylic acid, dimethyl ester, 1r,2s-rel, 1,2-dimethyl 1r,2s-cyclopropane-1,2-dicarboxylate, 1r,2s-dimethyl cyclopropane-1,2-dicarboxylate, 1,2-cyclopropanedicarboxylic acid, dimethyl ester, 1r,2s, 1,2-cyclopropanedicarboxylicacid, 1,2-dimethyl ester, 1r,2s-rel
PubChem CID 642979
IUPAC Name dimethyl (1S,2R)-cyclopropane-1,2-dicarboxylate
SMILES COC(=O)C1CC1C(=O)OC

Specifications

Melting Point 34°C to 38°C
Color Colorless
Density 1.1500g/mL
Boiling Point 112.0°C (18.0 mmHg)
Flash Point 73°C
Infrared Spectrum Authentic
Assay Percent Range 97% min. (GC)
Packaging Glass bottle
Refractive Index 1.4445 to 1.4465
Quantity 25 g
Specific Gravity 1.15
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in all organic solvents
Formula Weight 158.16
Percent Purity 97+%
Physical Form Liquid
Chemical Name or Material Dimethyl cis-1,2-cyclopropanedicarboxylate
Show More Show Less
compliance-icons
Product Identifier
  • Dimethyl cis-1,2-cyclopropanedicarboxylate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dimethyl cis-1,2-cyclopropanedicarboxylate, 97+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.