missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl-β-cyclodextrin, methylated β-cyclodextrins, 96%, Thermo Scientific Chemicals
£62.11 - £196.98
Chemical Identifiers
CAS | 51166-71-3 |
---|---|
Molecular Formula | C56H98O35 |
Molecular Weight (g/mol) | 1331.4 |
MDL Number | MFCD00011616 |
InChI Key | QGKBSGBYSPTPKJ-UZMKXNTCSA-N |
Synonym | 2,6-di-o-methyl-beta-cyclodextrin, unii-5fjp73e35o, dimethyl-beta-cyclodextrin, heptakis 2,6-o-dimethyl beta-cyclodextrin, heptakis-2,6-di-o-methyl-beta-cyclodextrin, heptakis 2,6-di-o-methyl-beta-cyclodextrin, dm-beta-cyd, dimethyl, a-cyclodextrin, 2,6-dme-beta-cyclodextrin, 2,6-di-o-methyl-beta-cd |
PubChem CID | 10171019 |
SMILES | COCC1C2C(C(C(O1)OC3C(OC(C(C3O)OC)OC4C(OC(C(C4O)OC)OC5C(OC(C(C5O)OC)OC6C(OC(C(C6O)OC)OC7C(OC(C(C7O)OC)OC8C(OC(O2)C(C8O)OC)COC)COC)COC)COC)COC)COC)OC)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10328230
|
Thermo Scientific Acros
235530010 |
1 g |
£62.11
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10045413
|
Thermo Scientific Acros
235530050 |
5 g |
£196.98
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagentChemical Identifiers
51166-71-3 | |
1331.4 | |
QGKBSGBYSPTPKJ-UZMKXNTCSA-N | |
10171019 |
C56H98O35 | |
MFCD00011616 | |
2,6-di-o-methyl-beta-cyclodextrin, unii-5fjp73e35o, dimethyl-beta-cyclodextrin, heptakis 2,6-o-dimethyl beta-cyclodextrin, heptakis-2,6-di-o-methyl-beta-cyclodextrin, heptakis 2,6-di-o-methyl-beta-cyclodextrin, dm-beta-cyd, dimethyl, a-cyclodextrin, 2,6-dme-beta-cyclodextrin, 2,6-di-o-methyl-beta-cd | |
COCC1C2C(C(C(O1)OC3C(OC(C(C3O)OC)OC4C(OC(C(C4O)OC)OC5C(OC(C(C5O)OC)OC6C(OC(C(C6O)OC)OC7C(OC(C(C7O)OC)OC8C(OC(O2)C(C8O)OC)COC)COC)COC)COC)COC)COC)OC)O |
Specifications
51166-71-3 | |
White to Yellow | |
(on dry basis) calculated | |
C56H98O35 | |
+163° (20°C c=1,water) | |
2,6-di-o-methyl-beta-cyclodextrin, unii-5fjp73e35o, dimethyl-beta-cyclodextrin, heptakis 2,6-o-dimethyl beta-cyclodextrin, heptakis-2,6-di-o-methyl-beta-cyclodextrin, heptakis 2,6-di-o-methyl-beta-cyclodextrin, dm-beta-cyd, dimethyl, a-cyclodextrin, 2,6-dme-beta-cyclodextrin, 2,6-di-o-methyl-beta-cd | |
QGKBSGBYSPTPKJ-UZMKXNTCSA-N | |
1331.4 | |
1331.4 | |
Crystalline Powder |
298.0°C to 310.0°C | |
Authentic | |
Glass bottle | |
MFCD00011616 | |
+ 163.00 | |
Solubility in water: >330g/l (25°C). Other solubilities: <10g/l boiling water,>250g/l methanol,chloroform,dmso,dmf and,methylene chloride (25°C),<50g/l acetone (25°C) | |
COCC1C2C(C(C(O1)OC3C(OC(C(C3O)OC)OC4C(OC(C(C4O)OC)OC5C(OC(C(C5O)OC)OC6C(OC(C(C6O)OC)OC7C(OC(C(C7O)OC)OC8C(OC(O2)C(C8O)OC)COC)COC)COC)COC)COC)COC)OC)O | |
10171019 | |
96% | |
Dimethyl-β-cyclodextrin, methylated β-cyclodextrins |
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title