missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl fumarate, 99%, Thermo Scientific Chemicals

Product Code. 11324489
Click to view available options
Quantity:
25 g
100 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Dimethyl fumarate acts as an immunomodulator. It is also used in cycloaddition reactions involving ylides, benzenes and amino acids. It is added to food grains,tobacco,leather and preservation. Clothing

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl fumarate acts as an immunomodulator. It is also used in cycloaddition reactions involving ylides, benzenes and amino acids. It is added to food grains,tobacco,leather and preservation. Clothing

Solubility
Soluble in water (1.6 mg/ml at 20°C), methanol (30-36 mg/ml), ethanol (10 mg/ml at 25°C), DMSO (29 mg/ml at 25°C), and DMF (∼12 mg/ml).

Notes
Do not store together with acids. Incompatible with acids, bases, oxidizing agents, reducing agents. Store at room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 624-49-7
Molecular Formula C6H8O4
Molecular Weight (g/mol) 144.126
MDL Number MFCD00064438
InChI Key LDCRTTXIJACKKU-ONEGZZNKSA-N
Synonym dimethyl fumarate, tecfidera, e-dimethyl fumarate, fumaderm, dimethyl e-but-2-enedioate, fumaric acid, dimethyl ester, boletic acid dimethyl ester, fumaric acid dimethyl ester, methyl fumarate, dimethyl trans-ethylenedicarboxylate
PubChem CID 637568
ChEBI CHEBI:76004
IUPAC Name dimethyl (E)-but-2-enedioate
SMILES COC(=O)C=CC(=O)OC

Specifications

Melting Point 102°C to 105°C
Density 1.37
Boiling Point 192°C to 193°C
Quantity 25 g
Beilstein 774590
Merck Index 14,4287
Solubility Information Soluble in water (1.6mg/ml at 20°C),methanol (30-36mg/ml),ethanol (10mg/ml at 25°C),DMSO (29mg/ml at 25°C),and DMF (∽12mg/ml).
Formula Weight 144.13
Percent Purity 99%
Chemical Name or Material Dimethyl fumarate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'