Learn More
Dimethyl fumarate, 99%, Thermo Scientific Chemicals
£24.70 - £92.10
Chemical Identifiers
CAS | 624-49-7 |
---|---|
Molecular Formula | C6H8O4 |
Molecular Weight (g/mol) | 144.126 |
MDL Number | MFCD00064438 |
InChI Key | LDCRTTXIJACKKU-ONEGZZNKSA-N |
Synonym | dimethyl fumarate, tecfidera, e-dimethyl fumarate, fumaderm, dimethyl e-but-2-enedioate, fumaric acid, dimethyl ester, boletic acid dimethyl ester, fumaric acid dimethyl ester, methyl fumarate, dimethyl trans-ethylenedicarboxylate |
PubChem CID | 637568 |
ChEBI | CHEBI:76004 |
IUPAC Name | dimethyl (E)-but-2-enedioate |
SMILES | COC(=O)C=CC(=O)OC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11324489
|
Thermo Scientific Alfa Aesar
A10402.14 |
25 g |
£24.70
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11334489
|
Thermo Scientific Alfa Aesar
A10402.22 |
100 g |
£47.60
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11408593
|
Thermo Scientific Alfa Aesar
A10402.30 |
250 g |
£92.10
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Dimethyl fumarate acts as an immunomodulator. It is also used in cycloaddition reactions involving ylides, benzenes and amino acids. It is added to food grains,tobacco,leather and preservation. Clothing
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDimethyl fumarate acts as an immunomodulator. It is also used in cycloaddition reactions involving ylides, benzenes and amino acids. It is added to food grains,tobacco,leather and preservation. Clothing
Solubility
Soluble in water (1.6 mg/ml at 20°C), methanol (30-36 mg/ml), ethanol (10 mg/ml at 25°C), DMSO (29 mg/ml at 25°C), and DMF (∼12 mg/ml).
Notes
Do not store together with acids. Incompatible with acids, bases, oxidizing agents, reducing agents. Store at room temperature.
Chemical Identifiers
624-49-7 | |
144.126 | |
LDCRTTXIJACKKU-ONEGZZNKSA-N | |
637568 | |
dimethyl (E)-but-2-enedioate |
C6H8O4 | |
MFCD00064438 | |
dimethyl fumarate, tecfidera, e-dimethyl fumarate, fumaderm, dimethyl e-but-2-enedioate, fumaric acid, dimethyl ester, boletic acid dimethyl ester, fumaric acid dimethyl ester, methyl fumarate, dimethyl trans-ethylenedicarboxylate | |
CHEBI:76004 | |
COC(=O)C=CC(=O)OC |
Specifications
624-49-7 | |
1.37 | |
C6H8O4 | |
774590 | |
dimethyl fumarate, tecfidera, e-dimethyl fumarate, fumaderm, dimethyl e-but-2-enedioate, fumaric acid, dimethyl ester, boletic acid dimethyl ester, fumaric acid dimethyl ester, methyl fumarate, dimethyl trans-ethylenedicarboxylate | |
LDCRTTXIJACKKU-ONEGZZNKSA-N | |
dimethyl (E)-but-2-enedioate | |
637568 | |
144.13 | |
Dimethyl fumarate |
102°C to 105°C | |
192°C to 193°C | |
MFCD00064438 | |
14,4287 | |
Soluble in water (1.6mg/ml at 20°C),methanol (30-36mg/ml),ethanol (10mg/ml at 25°C),DMSO (29mg/ml at 25°C),and DMF (∽12mg/ml). | |
COC(=O)C=CC(=O)OC | |
144.126 | |
CHEBI:76004 | |
99% |
Safety and Handling
GHS H Statement
H312-H315-H319-H317-H335
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P312-P333+P313-P362-P501c
H312-H315-H317-H319-H335
missing translation for 'einecsNumber' : 210-849-0
missing translation for 'rtecsNumber' : EM6125000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only