missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl isopropenyl phosphate, 95%, Thermo Scientific™

Product Code. 11357109
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4185-82-4
Molecular Formula C5H11O4P
Molecular Weight (g/mol) 166.113
MDL Number MFCD09953485
InChI Key CLUBBNMVLUHMDR-UHFFFAOYSA-N
Synonym dimethyl isopropenyl phosphate, phosphoric acid, isopropenyl dimethyl ester, phosphoric acid, dimethyl 1-methylethenyl ester, isopropenyl dimethyl phosphate, 1-propen-2-yl-dimethyl phosphate, dimethyl 1-methylethenyl phosphate, phosphoric acid dimethyl=1-methylvinyl ester, isopropenyl methyl phosphate, c3h5o meo 2po
PubChem CID 581450
IUPAC Name dimethyl prop-1-en-2-yl phosphate
SMILES CC(=C)OP(=O)(OC)OC

Specifications

Boiling Point 120°C (7.5 mmHg)
Refractive Index 1.42
Quantity 5 g
Sensitivity Moisture sensitive
Formula Weight 166.11
Percent Purity 95%
Chemical Name or Material Dimethyl isopropenyl phosphate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'