missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl selenide, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
Used as primary and secondary intermediate and also used in chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Color | Colorless |
Density | 1.41 g/mL |
Boiling Point | 56°C to 58°C |
Odor | Unpleasant |
Linear Formula | (CH3)2Se |
Quantity | 1 g |
UN Number | UN3282 |
Beilstein | 1696848 |
Sensitivity | Air and light sensitive |
Solubility Information | Very soluble in ethyl ether and ethyl alcohol. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'