missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl succinate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
250 g
1000 g
Description
Dimethyl succinate is used as a flavoring agent. It have wide range of industrial applications such as Functional fluids (open systems), Intermediates, Paint additives and coating additives, Pigments Solvents, Viscosity adjustors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Dimethyl succinate is used as a flavoring agent. It have wide range of industrial applications such as Functional fluids (open systems), Intermediates, Paint additives and coating additives, Pigments Solvents, Viscosity adjustors.
Solubility
Soluble in water. (122,9 g/L at 20°C). soluble in acetone, ethanol, very soluble in ethyl ether.
Notes
Store at room temperature. Ensure good ventilation.
Dimethyl succinate is used as a flavoring agent. It have wide range of industrial applications such as Functional fluids (open systems), Intermediates, Paint additives and coating additives, Pigments Solvents, Viscosity adjustors.
Solubility
Soluble in water. (122,9 g/L at 20°C). soluble in acetone, ethanol, very soluble in ethyl ether.
Notes
Store at room temperature. Ensure good ventilation.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 18°C to 19°C |
Density | 1.119 |
Boiling Point | 196°C |
Flash Point | 85°C (185°F) |
Refractive Index | 1.419 |
Quantity | 250 g |
Beilstein | 956776 |
Merck Index | 14,8869 |
Solubility Information | Soluble in water. (122,9g/L at 20°C). soluble in acetone,ethanol,very soluble in ethyl ether. |
Formula Weight | 146.14 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'